Learn More
Cobalt bis(trifluoromethylsulfonyl)imide, Thermo Scientific™
129.00€ - 525.00€
Chemical Identifiers
| CAS | 207861-61-8 |
|---|---|
| Fórmula molecular | C4CoF12N2O8S4 |
| Peso molecular (g/mol) | 619.204 |
| Número MDL | MFCD23380169 |
| Clave InChI | HRURXKIZWNSHQB-UHFFFAOYSA-N |
| Sinónimo | cobalt bis-trifluoromethanesulfonimide |
| PubChem CID | 131875824 |
| Nombre IUPAC | bis(trifluoromethylsulfonyl)azanide;cobalt(2+) |
| SMILES | C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2] |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|
15432837
|
Thermo Scientific Alfa Aesar
047053.03 |
1g |
129.00€
1 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
15442837
|
Thermo Scientific Alfa Aesar
047053.06 |
525.00€
5 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 207861-61-8 | |
| 619.204 | |
| HRURXKIZWNSHQB-UHFFFAOYSA-N | |
| 131875824 | |
| C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2] |
| C4CoF12N2O8S4 | |
| MFCD23380169 | |
| cobalt bis-trifluoromethanesulfonimide | |
| bis(trifluoromethylsulfonyl)azanide;cobalt(2+) |
Specifications
| Cobalt bis(trifluoromethylsulfonyl)imide | |
| C4CoF12N2O8S4 | |
| MFCD23380169 | |
| cobalt bis-trifluoromethanesulfonimide | |
| HRURXKIZWNSHQB-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;cobalt(2+) | |
| 131875824 | |
| Hygroscopic |
| 207861-61-8 | |
| [(CF3SO2)2N]2Co | |
| UN3261 | |
| It is soluble in water. | |
| C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Co+2] | |
| 619.204 | |
| 619.23 |
Safety and Handling
GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen