missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT10 Antibody (PCRP-CNOT10-1D5), mFluor Violet 450 SE, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
CNOT10 Monoclonal antibody specifically detects CNOT10 in Human samples. It is validated for Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | CNOT10 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | PCRP-CNOT10-1D5 |
| Conjugado | mFluor Violet 450 SE |
| Dilución | Flow Cytometry |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CCR4-NOT transcription complex subunit 10, CCR4-NOT transcription complex, subunit 10, DKFZp434K115, FLJ12890, FLJ13165 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant full-length human CNOT10 protein (Uniprot: Q9H9A5) |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?