Learn More
Cleavage stimulation factor 2 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™
Código de producto. 30514988 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30514988 0.1 mL 0.10 ml
1 options
Código de producto. 30514988 Proveedor Novus Biologicals N.º de proveedor NBP338509AF405
Rabbit Polyclonal Antibody
Cleavage stimulation factor 2 Polyclonal antibody specifically detects Cleavage stimulation factor 2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Cleavage stimulation factor 2
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Alexa Fluor 405
Formulación 50mM Sodium Borate
Alias de gen betaCstF-64 variant 2, CF-1 64 kDa subunit, Cleavage stimulation factor 64 kDa subunit, cleavage stimulation factor subunit 2, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kD, cleavage stimulation factor, 3' pre-RNA, subunit 2, 64kDa, CSTF 64 kDa subunit, CstF-64
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human Cleavage stimulation factor 2 (NP_001316.1).,, Sequence:, MAGLTVRDPAVDRSLRSVFVGNIPYEATEEQLKDIFSEVGPVVSFRLVYDRETGKPKGYGFCEYQDQETALSAMRNLNGREFSGRALRVDNAASEKNKEELKSLGTGAPVIESPYGETISPEDAPESISKAVASLPPEQMFELMKQMKLCVQNSPQEARNMLLQNPQLAYALLQAQVVMR
Método de purificación Affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1478
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Tipo de producto Antibody
Formulario Purified
Isotype IgG