missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Antibody (CGA/414), Novus Biologicals™

Código de producto. 18461962 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
20 ug
Tamaño de la unidad:
0.10 miligramo
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18461962 20 ug 20 microgramos
18017813 0.1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18461962 Proveedor Novus Biologicals N.º de proveedor NBP22942820ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody has been used in 4 publications

Chromogranin A Monoclonal specifically detects Chromogranin A in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Chromogranin A
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Clasificación Monoclonal
Clon CGA/414
Concentración 0.2 mg/ml
Conjugado Unconjugated
Dilución Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunoprecipitation 1-2ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml
N.º de referencia del gen P10645
Alias de gen betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I
Símbolos de los genes CHGA
Especie del huésped Mouse
Inmunógeno Recombinant full-length human Chromogranin A protein (Uniprot: P10645)
Método de purificación Protein A or G purified
Cantidad 20 ug
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Neuronal Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 1113
Especificidad de la prueba Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.
Especies diana Human
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.