Learn More
Cephalexin hydrate, 97+%, Thermo Scientific™
Cephalexin hydrate, cefalexin hydrate, CAS # 23325-78-2, is a first-generation cephalosporin antibiotic from the beta-lactam family.
Marca: Thermo Scientific Alfa Aesar J63172.14
| Cantidad | 25g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Cephalexin hydrate is the hydrate of the cephalosporin antibiotic, cephalexin
• Cephalexin is a beta-lactam antibiotic that inhibits bacterial growth and division
• Cephalexin binds to penicillin-binding protein and inhibits cell wall synthesis
Application
• Cephalexin works best on gram-positive microbes
• Useful for immunology research, particularly on antibiotic allergies and cross-reactivity
| 15686-71-2 | |
| 401.862 | |
| YHJDZIQOCSDIQU-OEDJVVDHSA-N | |
| (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate |
| C16H20ClN3O5S | |
| MFCD00167148 | |
| 57515946 | |
| [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-] |
Especificaciones
| Cephalexin hydrate | |
| White | |
| C16H20ClN3O5S | |
| 965503 | |
| Soluble in DMSO and Methanol. | |
| [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-] | |
| 401.862 | |
| 347.39 (Anhydrous) |
| 15686-71-2 | |
| 25g | |
| MFCD00167148 | |
| 141974 | |
| YHJDZIQOCSDIQU-OEDJVVDHSA-N | |
| (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate | |
| 57515946 | |
| ≥97% |