Learn More
Cefixime trihydrate, Thermo Scientific™
A broad spectrum third generation cephalosporin
Marca: Thermo Scientific Alfa Aesar J66094.03
| Cantidad | 1g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 125110-14-7 | |
| 453.44 | |
| OKBVVJOGVLARMR-QSWIMTSFSA-N | |
| CHEBI:472657 | |
| NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O |
| C16H15N5O7S2 | |
| MFCD00865020 | |
| 5362065 | |
| (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid |
Especificaciones
| Cefixime trihydrate | |
| Yellow | |
| Air and light sensitive | |
| MFCD00865020 | |
| OKBVVJOGVLARMR-QSWIMTSFSA-N | |
| (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[(carboxymethoxy)imino]acetamido]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid | |
| 5362065 | |
| 507.49 |
| 125110-14-7 | |
| 1g | |
| C16H15N5O7S2 | |
| Soluble in water at 55;1 μg/mL | |
| NC1=NC(=CS1)C(=N\OCC(O)=O)\C(=O)N[C@H]1[C@H]2SCC(C=C)=C(N2C1=O)C(O)=O | |
| 453.44 | |
| CHEBI:472657 |