Learn More
Eurofins DiscoverX Cdk2/Cyclin E Protein, Active
Descripción
The cyclin-dependent kinases (CDKs), are a family of serine threonine protein kinases, that control the cell division cycle. In mammals, nine different CDKs (cdk 1 to cdk 9) have been identified. CDKs are regulated on various levels including binding of proteins (cyclins, inhibitors, and assembly factors), protein degradation, transcriptional control, localization, and multiple phosphorylations. This leads to timely regulation of CDK activity, allowing progression from one cell cycle phase to the next. The cdk2 promotes the G1/S trasition and progression through the S phase when bound to specific cyclins (E and A). Recent evidences indicate that CDK2 is involved in cell cycle independent functions such as DNA damage repair.
EMBL M68520 / GenBank NM_001238
Especificaciones
Especificaciones
| Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol, 270 mM sucrose. Frozen solution. |
| Peso molecular | 34 kDa (CDK2) and 74 kDa (Cyclin E1) |
| Cantidad | 10 μg |
| Especie | Human |
| Recombinante | Recombinant |
| Etiqueta de proteína | C-terminal 6His / N-terminal GST |
| Sistema de expresión | Recombinant enzyme expressed in Sf21 Insect cells |
| Forma de proteína | Full-length |
| Grado de pureza o calidad | Combined ≥51% |
| Expresión | CDK2/Cyclin E, active |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.