Learn More
CCL4/MIP-1 beta Antibody - Azide and BSA Free, Novus Biologicals™
Código de producto. 18606856 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18606856 100 μg 100 microlitros
18603948 20 μg 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18606856 Proveedor Novus Biologicals N.º de proveedor NBP305561100ul
Rabbit Polyclonal Antibody
CCL4/MIP-1 beta Polyclonal antibody specifically detects CCL4/MIP-1 beta in Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CCL4/MIP-1 beta
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500 - 1:1000
Formulación PBS with 50% glycerol, pH7.3.
Alias de gen ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to a sequence within amino acids 1-92 of human CCL4/MIP-1 beta (NP_002975.1). MKLCVTVLSLLMLVAAFCSPALSAPMGSDPPTACCFSYTARKLPRNFVVDYYETSSLCSQPAVVFQTKRSKQVCADPSESWVQEYVYDLELN
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Biologically Active Proteins, Chemokines and Cytokines
Primario o secundario Primary
ID de gen (Entrez) 6351
Especies diana Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG