missing translation for 'onlineSavingsMsg'
Learn More

CCL14/HCC-1/HCC-3 Antibody (015), DyLight 488, R&D Systems™

Código de producto. 18498695 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Tamaño de la unidad
18498695 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18498695 Proveedor Novus Biologicals N.º de proveedor NBP289521G
 Artículo restringido
Fecha estimada de envoi: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

CCL14/HCC-1/HCC-3 Monoclonal specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CCL14/HCC-1/HCC-3
Aplicaciones ELISA
Clasificación Monoclonal
Clon 015
Conjugado DyLight 488
Dilución ELISA
Formulación 50mM Sodium Borate
Alias de gen C-C motif chemokine 14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14
Especie del huésped Rabbit
Inmunógeno This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL14/HCC-1/HCC-3 (Q16627-1; Thr20-Asn93).
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primario o secundario Primary
ID de gen (Entrez) 6358
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Tipo de producto Antibody
Formulario Purified
Isotype IgG
Línea de productos Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.