Learn More
Carbonic Anhydrase IX/CA9 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18785623 Tienda Bio Techne Productos
Cambiar vista
Click to view available options
Cantidad:
0.025 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18785623 0.025 mL 25 microlitros
18797463 - 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18785623 Proveedor Novus Biologicals N.º de proveedor NB100417SS
Rabbit Polyclonal Antibody
Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Mouse, Plant, Rat samples. It is validated for Chemotaxis, ChIP Assay, Dual RNAScope ISH-IHC, ELISA, EMSA, Flow Cytometry, Gel Super Shift Assays, IF/ICC, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Microarray, Proximity Ligation Assay, Simple Western, Westen Blot, Western Blot.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Carbonic Anhydrase IX/CA9
Aplicaciones Chemotaxis, ChIP Assay, Dual RNAScope ISH-IHC, ELISA, EMSA, Flow Cytometry, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Microarray, Proximity Ligation Assay, Simple Western, Western Blot, Western Blot
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 - 3 ug/ml, Simple Western 1:50, Flow Cytometry 1:1000, ELISA reported in scientific literature (PMID 19963243), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation 1:10 - 1:500. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray reported in scientific literature (PMID 31955345), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Dual RNAscope ISH-IHC 1:1000
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen Q16790
Alias de gen CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Símbolos de los genes CA9
Especie del huésped Rabbit
Inmunógeno A synthetic peptide made to a C-terminal sequence of human Carbonic Anhydrase IX (within residues 400-459) [UniProt Q16790]
Peso molecular del antígeno 55 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 768
Especificidad de la prueba Human, dog, rat and mouse (PMID 22842475).
Especies diana Human, Mouse, Mouse, Plant, Rat
Contenido y almacenamiento Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only