missing translation for 'onlineSavingsMsg'
Learn More

Calmodulin Antibody (2D1), Alexa Fluor™ 532, Novus Biologicals™

Código de producto. 18714763 Tienda Bio Techne Productos
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
0.2 mL
missing translation for 'unitSize'
0.20 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18714763 0.2 mL 0.20 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18714763 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NB1202860AF532

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calmodulin Monoclonal antibody specifically detects Calmodulin in Human, Mouse, Rat, Bacteria, Bovine, Chicken samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Calmodulin
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 2D1
Conjugado Alexa Fluor 532
Dilución Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulación 50mM Sodium Borate
Alias de gen CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit
Especie del huésped Mouse
Inmunógeno Calmodulin purified from Dictyostelium discoideum.
Método de purificación Protein A purified
Cantidad 0.2 mL
Estado normativo RUO
Disciplina de investigación Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 801
Especies diana Human, Mouse, Rat, Bacteria, Bovine, Chicken
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.