missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calbindin D-28K Antibody (CL15065) [Janelia Fluor™ 549], Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Calbindin D-28K Monoclonal antibody specifically detects Calbindin D-28K in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Calbindin D-28K |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CL15065 |
| Conjugado | Janelia Fluor 549 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CAB27, CALB, calbindin, calbindin 1, (28kD), calbindin 1, 28kDa, Calbindin D28, D-28K, RTVL-H protein, Vitamin D-dependent calcium-binding protein, avian-type |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was generated using a recombinant protein sequence of P05937, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?