missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
CACNB1 Monoclonal antibody specifically detects CACNB1 in Human,Mouse samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | CACNB1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI3H6 |
| Conjugado | Janelia Fluor 525 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | CAB1, CACNLB1calcium channel, L type, beta 1 polypeptide, Calcium channel voltage-dependent subunit beta 1, calcium channel, voltage-dependent, beta 1 subunit, CCHLB1, dihydropyridine-sensitive L-type, calcium channel beta-1 subunit, MGC41896, voltage-dependent L-type calcium channel subunit beta-1 |
| Especie del huésped | Mouse |
| Inmunógeno | Human recombinant protein fragment corresponding to amino acids 1-269 of human CACNB1(NP_954855) produced in E.coli. |
| Método de purificación | >95% |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?