Learn More
Biotin (Vitamin B7 or Vitamin H) Mouse anti-All species, Clone: BTN/36, Novus Biologicals™
Código de producto. 18304105 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18304105 100 μg 100 microgramos
18328661 20 μg 20 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18304105 Proveedor Novus Biologicals N.º de proveedor NBP313907100UG
Mouse Monoclonal Antibody
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in All species samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Biotin (Vitamin B7 or Vitamin H)
Aplicaciones Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH)
Clasificación Monoclonal
Clon BTN/36
Concentración 0.2 mg/ml
Conjugado Unconjugated
Dilución Western Blot 0.5-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin 0.5-1 ug/ml, In Situ Hybridization (ISH)
Formulación 10 mM PBS with 0.05% BSA
Alias de gen 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H
Especie del huésped Mouse
Inmunógeno Biotinylated sheep immunoglobulin
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana All species
Contenido y almacenamiento Store at 4°C.
Formulario Purified
Isotype IgG1 κ