Learn More
BI-882370, MedChemExpress
Código de producto. 16465116
Encompass_Preferred
Cambiar vista
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Tamaño de la unidad:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
16465116 100 mg 100 miligramos
16434158 10 mg 10 miligramos
16430507 50 mg 50 miligramos
16416867 5 mg 5 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Código de producto. 16465116 Proveedor Medchem Express N.º de proveedor HY107779
MedChemExpress BI-882370 is a potent and selective RAF kinase inhibitor that binds to the ATP binding site of the kinase positioned in the DFG-out (inactive) conformation of the BRAF kinase. BI-882370 (BI 882370) inhibits the oncogenic BRAFV600E-mutant, the WT BRAF and CRAF kinases with IC50s of 0.4, 0.8, and 0.6 nM, respectively. BI-882370 also inhibits SRC family kinases.
BI-882370 is a potent and selective RAF kinase inhibitor that binds to the ATP binding site of the kinase positioned in the DFG-out (inactive) conformation of the BRAF kinase. BI-882370 (BI 882370) inhibits the oncogenic BRAFV600E-mutant, the WT BRAF and CRAF kinases with IC50s of 0.4, 0.8, and 0.6 nM, respectively. BI-882370 also inhibits SRC family kinases.
Especificaciones
Nombre del producto químico o material BI-882370
CAS 1392429-79-6
Color Caqui
Notas de grado de pureza Research
Fórmula molecular C28H33F2N7O2S
Cantidad 100 mg
Información de solubilidad DMSO : 5 mg/mL (8.78 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CCCS(=O)(NC1=CC=C(F)C(N2C=C(C3=CN=CN=C3)C4=NC(N(C5CCN(CC)CC5)C)=CC=C42)=C1F)=O
Peso molecular (g/mol) 569.67
Formula Weight (peso de la fórmula) 569.67
Porcentaje de pureza 95.02%
Grado Research
Forma física Solid
Para utilizar con (aplicación) Cancer-Kinase/protease