Learn More
beta-III Tubulin Antibody (CL5813) [Alexa Fluor™ 594], Novus Biologicals™
Código de producto. 30294243 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30294243 0.1 mL 0.10 ml
1 options
Código de producto. 30294243 Proveedor Novus Biologicals N.º de proveedor NBP261430AF594
Mouse Monoclonal Antibody
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno beta-III Tubulin
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL5813
Conjugado Alexa Fluor 594
Formulación 50mM Sodium Borate
Alias de gen beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from Q13509, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 10381
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1