Learn More
Berkshire International BCR™ Aluminium Cleanroom Flat Mops with Springs
Código de producto. 18824741 Tienda Berkshire Productos
Change view
Click to view available options
Material:
Anodized Aluminum
Código de producto. Material
18824741 Anodized Aluminum
1 options
Código de producto. 18824741 Proveedor Berkshire International N.º de proveedor BCR.MOP2.10
Flat Mop Head are light and easy to maneuver, even with wet wipers attached.
  • Clean, light and durable
  • High quality anodized aluminum
  • Autoclavable
  • Unique stainless steel spring mechanisms for wiper attachment (BCR.MOP2.10)
Especificaciones
Tipo de producto Cleanroom Mop
Esterilidad Non-sterile
Esterilizables en autoclave Autoclavable
Desechables Reusable
Color-coded No
Material Anodized Aluminum