Description
- Clean, light and durable
- High quality anodized aluminum
- Autoclavable
- Unique stainless steel spring mechanisms for wiper attachment (BCR.MOP2.1)
Specifications
Specifications
| Tipo de producto | Cleanroom Mop |
| Esterilidad | Non-sterile |
| Esterilizables en autoclave | Autoclavable |
| Desechables | Reusable |
| Color-coded | No |
| Material | Anodized Aluminum |