Learn More
Berkshire International BCR™ Aluminium Cleanroom Flat Mops
Código de producto. 18803617 Tienda Berkshire Productos
Change view
Click to view available options
Material:
Anodized Aluminum
Unit Size:
cada uno
Product Code. Material unitSize
18803617 Anodized Aluminum cada uno
1 options
This item is not returnable. View return policy
Product Code. 18803617 Supplier Berkshire International Supplier No. BCR.MOP2.1
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Flat Mop Head are light and easy to maneuver, even with wet wipers attached.
  • Clean, light and durable
  • High quality anodized aluminum
  • Autoclavable
  • Unique stainless steel spring mechanisms for wiper attachment (BCR.MOP2.1)
Specifications
Tipo de producto Cleanroom Mop
Esterilidad Non-sterile
Esterilizables en autoclave Autoclavable
Desechables Reusable
Color-coded No
Material Anodized Aluminum