Learn More
Isofield™ Atto 230 Cleanroom Irradiated Tie-On Facemask with Anti Fog Visor - Grade A/B Compatible
Descripción
- Irradiated tie-on cleanroom facemask with anti-fog visor
- 3-layer non-woven configuration
- Threaded top ties to ensure secure fit to face
- Anti-fog-coated visor
- Highly efficient bacterial and particle filtration
- Particle Shedding (Helmke Drum) Category I
- Individually packed pouches printed with IPA resistant ink to maintain traceability
Especificaciones
Especificaciones
| Tipo de producto | Cleanroom Face Mask |
| Color | Blanco |
| Fastening Type | Lazos |
| Material | Polyethylene, Polypropylene |
| Tamaño | Wide |
| Esterilidad | Estéril |
| Modelo | Plisado |