missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
ATG3/APG3 Monoclonal antibody specifically detects ATG3/APG3 in Canine,Human,Monkey,Mouse,Primate,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | ATG3/APG3 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | OTI3H2 |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | APG3, APG3 autophagy 3-like (S. cerevisiae), APG3L, APG3-LIKE, ATG3 autophagy related 3 homolog (S. cerevisiae), Autophagy-related protein 3, DKFZp564M1178, EC 6.3.2.-, FLJ22125,2610016C12Rik, hApg3, MGC15201, PC3-96, Protein PC3-96, ubiquitin-like-conjugating enzyme ATG3 |
| Especie del huésped | Mouse |
| Inmunógeno | Full length human recombinant protein of human ATG3(NP_071933) produced in HEK293T cell. |
| Método de purificación | Immunogen affinity purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?