missing translation for 'onlineSavingsMsg'
Learn More

APE Antibody - BSA Free, Novus Biologicals™

Código de producto. 18726723 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18726723 0.025 mL 25 microlitros
18779623 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18726723 Proveedor Novus Biologicals N.º de proveedor NB100101SS

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 64 publications

APE Polyclonal specifically detects APE in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno APE
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:1000, Simple Western 1:12.5, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 7 ug/ml, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Immunohistochemistry Free-Floating reported in scientific literature (PMID 17332344), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize
Formulación Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
N.º de referencia del gen P27695
Alias de gen AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease
Símbolos de los genes APEX1
Especie del huésped Rabbit
Inmunógeno Affinity purified human APE1 [UniProt# P27695]
Peso molecular del antígeno 37 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 328
Especificidad de la prueba Human, mouse, primate and rat. Rabbit reactivity reported in the scientific literature (PMID: 15276530).
Especies diana Human, Mouse, Rat, Primate, Rabbit
Contenido y almacenamiento Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.