missing translation for 'onlineSavingsMsg'
Learn More

AIMP2 Antibody, Novus Biologicals™

Código de producto. 18450061 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25 μL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18450061 25 μL 25 microlitros
18287116 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18450061 Proveedor Novus Biologicals N.º de proveedor NBP18157425ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AIMP2 Polyclonal specifically detects AIMP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno AIMP2
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
N.º de referencia del gen Q13155
Alias de gen aminoacyl tRNA synthase complex-interacting multifunctional protein 2, aminoacyl tRNA synthetase complex-interacting multifunctional protein 2, ARS-interacting multi-functional protein 2, JTV1JTV-1, Multisynthase complex auxiliary component p38, multisynthetase complex auxiliary component p38, p38, PRO0992, Protein JTV-1
Símbolos de los genes AIMP2
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:SVLGKDYGALKDIVINANPASPPLSLLVLHRLLCEHFRVLSTVHTHSSVKSVPENLLKCFGEQNKKQPRQDYQLGFTLIWKNVPKTQ
Peso molecular del antígeno 35 kDa
Método de purificación Affinity Purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7965
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.