missing translation for 'onlineSavingsMsg'
Learn More

ABCA6 Antibody - Azide and BSA Free, Novus Biologicals™

Código de producto. 18696962 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mL
0.1 mL
Tamaño de la unidad:
0.02 ml
0.10 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18696962 0.02 mL 0.02 ml
18682282 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18696962 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP2924970.02ml

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCA6 Polyclonal antibody specifically detects ABCA6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ABCA6
Aplicaciones Western Blot, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Formulación PBS with 50% glycerol, pH7.3.
Alias de gen ABC transporter ABCA6, ATP-binding cassette A6, ATP-binding cassette sub-family A member 6, ATP-binding cassette, sub-family A (ABC1), member 6, EC 2.5.1.19, EC 3.6.3, EC 3.6.3.25, EST155051, FLJ43498
Especie del huésped Rabbit
Inmunógeno Recombinant fusion protein containing a sequence corresponding to amino acids 690-828 of human ABCA6 (NP_525023.2). RLKCAGSSMFLKRRWGLGYHLSLHRNEICNPEQITSFITHHIPDAKLKTENKEKLVYTLPLERTNTFPDLFSDLDKCSDQGVTGYDISMSTLNEVFMKLEGQSTIEQDFEQVEMIRDSESLNEMELAHSSFSEMQTAVS
Método de purificación Affinity purified
Cantidad 0.02 mL
Estado normativo RUO
Disciplina de investigación Cardiovascular Biology, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 23460
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.