missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
AAR2 Polyclonal antibody specifically detects AAR2 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antígeno | AAR2 |
| Aplicaciones | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | AAR2 splicing factor homolog (S. cerevisiae), bA234K24.2, C20orf4, CGI-23, chromosome 20 open reading frame 4, DKFZp564N1363, hypothetical protein LOC25980 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human AAR2 (NP_056326.2).,, Sequence:, MAAVQMDPELAKRLFFEGATVVILNMPKGTEFGIDYNSWEVGPKFRGVKMIPPGIHFLHYSSVDKANPKEVGPRMGFFLSLHQRGLTVLRWSTLREEVDLSPAPESEVEAMRANLQELDQFLGPYPYATLKKWISLTNFISEATVEKLQPENRQICAFSDVLPVLSMKHTKDRVGQNLPRCGIECKSYQEGLARLPEMKPRAGTEIRFSELPTQMFPEGATPAEITKHSMDLSYALETVLNKQFPSSPQD |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?