missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AADACL4 Polyclonal specifically detects AADACL4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antígeno | AADACL4 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/ml |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1.0 ug/ml |
| Formulación | PBS, 2% Sucrose with 0.09% Sodium Azide |
| N.º de referencia del gen | NP_001013652 |
| Alias de gen | arylacetamide deacetylase-like 4 |
| Símbolos de los genes | AADACL4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?