missing translation for 'onlineSavingsMsg'
Learn More

2-Butynoic acid, 98%

Product Code. 11422977
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11422977

Brand: Thermo Scientific Alfa Aesar A15542.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-93-2 | C4H4O2 | 84.07 g/mol

2-Butynoic acid is used as an intermediate in organic synthesis and pharmaceuticals. It is used as a synthon in various reactions, which includes cylcoalkylation of phenols to flavones and chromones. It is involved in the cyclization of gamma-butyrolactones. It is also used in the synthesis of Z-trisubstituted olefins through μ-alkylation. It plays an important role in the preparation of chirally pure 1,2,3,4-tetrahydroisoquinoline analogs, which finds application as anti-cancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butynoic acid is used as an intermediate in organic synthesis and pharmaceuticals. It is used as a synthon in various reactions, which includes cylcoalkylation of phenols to flavones and chromones. It is involved in the cyclization of gamma-butyrolactones. It is also used in the synthesis of Z-trisubstituted olefins through µ-alkylation. It plays an important role in the preparation of chirally pure 1,2,3,4-tetrahydroisoquinoline analogs, which finds application as anti-cancer agents.

Solubility
Soluble in water. Slightly soluble in methanol and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-93-2
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00004363
InChI Key LUEHNHVFDCZTGL-UHFFFAOYSA-N
Synonym 2-butynoic acid, tetrolic acid, 3-methylpropiolic acid, 1-propynecarboxylic acid, 2-butynoicacid, tetrolsaure, butynoic acid, 2-butynic acid, propinecarboxylic acid
PubChem CID 68535
IUPAC Name but-2-ynoic acid
SMILES CC#CC(O)=O

Specifications

Melting Point 75°C to 79°C
Density 0.964
Boiling Point 200°C to 203°C
Linear Formula CH3C≡CCO2H
Quantity 25 g
UN Number UN3261
Beilstein 1740205
Solubility Information Soluble in water. Slightly soluble in methanol and chloroform.
Formula Weight 84.07
Percent Purity 98%
Chemical Name or Material 2-Butynoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.