missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
14-3-3 eta Monoclonal specifically detects 14-3-3 eta in Bovine, Canine, Equine, Human, Mouse, Rat samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.
Specifications
Specifications
| Aplicaciones | Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot |
| Conjugado | DyLight 650 |
| Dilución | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Knockout Validated |
| Método de purificación | Immunogen affinity purified |
| Cantidad | 0.1 mL |
| Estado normativo | RUO |
| Especies diana | Bovine, Canine, Equine, Human, Mouse, Rat |
| Contenido y almacenamiento | Store at 4C in the dark. |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?