Learn More
1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%, Thermo Scientific™
Marca: Thermo Scientific Alfa Aesar H36633.03
| Cantidad | 1g |
|---|
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
| 286014-38-8 | |
| 320.18 | |
| CQHXJIHJFMBBQA-UHFFFAOYSA-N | |
| 16218169 | |
| F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 |
| C15H25BF4N2 | |
| MFCD04973310 | |
| 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt | |
| 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide |
Especificaciones
| 1,3-Dicyclohexylimidazolium tetrafluoroborate | |
| C15H25BF4N2 | |
| 1g | |
| 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt | |
| F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1 | |
| 320.18 | |
| 320.18 |
| 286014-38-8 | |
| MFCD04973310 | |
| Moisture sensitive | |
| CQHXJIHJFMBBQA-UHFFFAOYSA-N | |
| 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide | |
| 16218169 | |
| 97% |