CAS RN 376594-67-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LY450108, MedChemExpress
MedChemExpress LY450108 is a potent AMPA receptor potentiator. LY450108 has the potential for depression and Parkinson's disease research.