CAS RN 959746-77-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
A-836339, MedChemExpress
MedChemExpress A-836339 is a cannabinoid CB2 receptor-selective agonist; exhibits high potencies at CB(2) and selectivity over CB(1) receptors.