CAS RN 957209-68-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NVP-QAV-572, MedChemExpress
MedChemExpress NVP-QAV-572 is a PI3K inhibitor extracted from patent US7998990B2, Compound Example 8, has an IC50 of 10 nM.