CAS RN 94424-50-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Quinagolide hydrochloride, MedChemExpress
MedChemExpress Quinagolide hydrochloride is a selective dopamine D2 receptor agonist, also is a prolactin inhibitor.