CAS RN 940310-85-0
IUPAC Nombre:
4-methyl-3-{[1-methyl-6-(pyridin-3-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl]amino}-N-[3-(trifluoromethyl)phenyl]benzamide
Sinónimos:
4-methyl-3-{[1-methyl-6-(pyridin-3-yl)pyrazolo[3,4-d]pyrimidin-4-yl]amino}-N-[3-(trifluoromethyl)phenyl]benzamide
Peso molecular (g/mol):
503.49
Fórmula molecular:
C26H20F3N7O
InChi Key:
ZCCPLJOKGAACRT-UHFFFAOYSA-N
SMILES:
CN1N=CC2=C(NC3=C(C)C=CC(=C3)C(=O)NC3=CC=CC(=C3)C(F)(F)F)N=C(N=C12)C1=CN=CC=C1
1
–
2
de
2
resultados
2
NVP-BHG712, MedChemExpress
MedChemExpress NVP-BHG712 is an oral active EphB4 kinase autophosphorylation inhibitor, with IC50 values of 3.3 nM and 3.0 nM for EphA2 and EphB4, respectively.