CAS RN 939055-18-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
FIPI, MedChemExpress
MedChemExpress FIPI is a derivative of halopemide which potently inhibits both PLD1 and PLD2 with IC50s of 25 nM and 20 nM, respectively.