CAS RN 938448-87-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bucladesine calcium, MedChemExpress
MedChemExpress Bucladesine calcium salt (Dibutyryl-cAMP calcium salt;DC2797 calcium salt) is a cell-permeable cyclic AMP (cAMP) analog and selectively activates cAMP dependent protein kinase (PKA) by increasing the intracellular level of cAMP. Bucladesine calcium salt acts as a phosphodiesterase (PDE) inhibitor.