CAS RN 935693-62-2
IUPAC Nombre:
N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine
Sinónimos:
N-(1-benzylpiperidin-4-yl)-6,7-dimethoxy-2-(4-methyl-1,4-diazepan-1-yl)quinazolin-4-amine
Peso molecular (g/mol):
490.65
Fórmula molecular:
C28H38N6O2
InChi Key:
OSXFATOLZGZLSK-UHFFFAOYSA-N
SMILES:
COC1=CC2=NC(=NC(NC3CCN(CC4=CC=CC=C4)CC3)=C2C=C1OC)N1CCCN(C)CC1
1
–
2
de
2
resultados
2
BIX-01294, MedChemExpress
MedChemExpress BIX-01294 is a reversible and highly selective G9a and GLP Histone Methyltransferase inhibitor, with IC50s of of 1.7 μM and 0.9 μM, respectively. BIX-01294 inhibits G9a/GLP by competing for binding with the amino acids N-terminal of the substrate lysine residue. BIX-01294, a (1H-1,4-diazepin-1-yl)-quinazolin-4-yl amine derivative, induces necroptosis and autophagy. BIX-01294 has antitumor activity in recurrent tumor cells.