CAS RN 935467-97-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TH-237A, MedChemExpress
MedChemExpress TH-237A(meso-GS 164) is a novel neuroprotective agent exhibiting favorable permeation across the blood brain barrier.