CAS RN 931706-15-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PPQ-102, MedChemExpress
MedChemExpress PPQ-102 is a potent CFTR inhibitor which can completely inhibited CFTR chloride current with IC50 of ∼90 nM.