CAS RN 92953-10-1
IUPAC Nombre:
[4-(4-chlorophenyl)butyl]diethylheptylazanium 4-methylbenzene-1-sulfonate
Sinónimos:
clofilium tosylate
Peso molecular (g/mol):
510.17
Fórmula molecular:
C28H44ClNO3S
InChi Key:
MOQZYUUHIWPDQC-UHFFFAOYSA-M
SMILES:
CC1=CC=C(C=C1)S([O-])(=O)=O.CCCCCCC[N+](CC)(CC)CCCCC1=CC=C(Cl)C=C1
1
–
1
de
1
resultados
1
Clofilium tosylate, MedChemExpress
MedChemExpress Clofilium tosylate, a potassium channel blocker, induces apoptosis of human promyelocytic leukemia (HL-60) cells via Bcl-2-insensitive activation of caspase-3. Antiarrhythmic agent.