CAS RN 926927-42-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Toll-like receptor modulator, MedChemExpress
MedChemExpress Toll-like receptor modulator is a modulator of TLR7/8, which modulates immune function.