CAS RN 92510-91-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
F 16915, MedChemExpress
MedChemExpress F 16915, a Docosahexaenoic Acid (DHA, HY-B2167) derivative, is a potent pro-drug of DHA. F 16915 can prevent heart failure-induced atrial fibrillation.