CAS RN 920022-47-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
RG7713, MedChemExpress
MedChemExpress RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a).