CAS RN 916603-07-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CZC-8004, MedChemExpress
MedChemExpress CZC-8004 is a pan-kinase inhibitor and binds a range of tyrosine kinases, including ABL kinase.