CAS RN 912445-05-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ Veliparib dihydrochloride
High affinity PARP-1 and -2 inhibitor; orally bioavailable
2
Veliparib dihydrochloride, MedChemExpress
MedChemExpress Veliparib (dihydrochloride) is a potent inhibitor of PARP1 and PARP2 with Kis of 5.2 nM and 2.9 nM in cell-free assays, respectively.