Learn More
Tocris Bioscience™ Veliparib dihydrochloride
Descripción
Veliparib dihydrochloride is a high affinity PARP-2 and PARP-1 inhibitor (Ki values are 2.9 and 5.2 nM, respectively). Inhibits repair of radiation-induced DNA damage and sensitizes lung cancer cells to radiation. Also increases autophagy and apoptosis in lung cancer cells. Enhances radiation-induced tumor growth delay in mice bearing lung tumor xenografts. Also chemosensitizer. Potentiates effects of temozolomide (Cat. No. 2706) in a mouse melanoma model. Orally bioavailable and brain penetrant.
Especificaciones
Especificaciones
| Nombre del producto químico o material | 2-[(2R)-2-Methyl-2-pyrrolidinyl]-1H-benzimidazole-4-carboxamide dihydrochloride |
| CAS | 912445-05-7 |
| Cantidad | 50 mg |
| Fórmula molecular | C13H16N4O.2HCl |
| Pureza | 98% |