CAS RN 911222-45-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ LY 2603618
Potent and selective Chk1 inhibitor (IC50 = 7 nM)
2
Rabusertib, MedChemExpress
MedChemExpress Rabusertib (LY2603618) is a potent and selective inhibitor of Chk1 with an IC50 of 7 nM.