CAS RN 910634-41-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Litronesib, MedChemExpress
MedChemExpress Litronesib (LY2523355) is a selective mitosis-specific kinesin Eg5 inhibitor, with antitumor activity.