CAS RN 908601-49-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SB-616234-A, MedChemExpress
MedChemExpress SB-616234-A is a selective and orally bioavailable 5-HT1B receptor antagonist, with anxiolytic and antidepressant activity.