CAS RN 9063-57-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Tuftsin
MedChemExpress Tuftsin is a tetrapeptide. Tuftsin is a macrophage/microglial activator.
2
MedChemExpress Tuftsin
MedChemExpress Tuftsin is a tetrapeptide. Tuftsin is a macrophage/microglial activator.
3
MedChemExpress Tuftsin
MedChemExpress Tuftsin is a tetrapeptide. Tuftsin is a macrophage/microglial activator.