CAS RN 901119-35-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Fostamatinib, TRC
CAS: 901119-35-5 Fórmula molecular: C23H26FN6O9P Peso molecular (g/mol): 580.47 Sinónimo: R 788,6-[[5-Fluoro-2-[(3,4,5-trimethoxyphenyl)amino]-4-pyrimidinyl]amino]-2,2-dimethyl-4-[(phosphonooxy)methyl]-2H-pyrido[3,2-b]-1,4-oxazin-3(4H)-one SMILES: COc1cc(Nc2ncc(F)c(Nc3ccc4OC(C)(C)C(=O)N(COP(=O)(O)O)c4n3)n2)cc(OC)c1OC
2
Fostamatinib, MedChemExpress
MedChemExpress Fostamatinib (R788) is the oral prodrug of the active compound R406. R406 is an orally available and competitive Syk/FLT3 inhibitor with a Ki of 30 nM and an IC50 of 41 nM. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).